Learn More
2,4'-Bipyridine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,4'-Bipyridine |
| CAS | 581-47-5 |
| Molecular Formula | C10H8N2 |
| Quantity | 5 g |
| Synonym | 2,4'-Bipyridyl, 2,4'-Dipyridine, 2,4'-Dipyridyl, 2-(Pyridin-4-yl)pyridine, 2-Pyridin-4-ylpyridine, 4-(2-Pyridyl)pyridine |
| InChI Formula | InChI=1S/C10H8N2/c1-2-6-12-10(3-1)9-4-7-11-8-5-9/h1-8H |
| SMILES | c1ccc(nc1)c2ccncc2 |
| IUPAC Name | 2-pyridin-4-ylpyridine |
| Molecular Weight (g/mol) | 156.18 |
| Formula Weight | 156.0687 |