missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Bipyridine, 97%

Product Code. 11367139
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11367139

Brand: Thermo Scientific Alfa Aesar H27366.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 581-47-5 | C10H8N2 | 156.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 581-47-5
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006217
InChI Key RMHQDKYZXJVCME-UHFFFAOYSA-N
Synonym 2,4'-bipyridine, 2,4'-dipyridyl, 4-pyridylpyridine, 2,4'-bipyridyl, 2,4-bipyridyl, 2,4'-dipyridine, unii-c6sn6ru851, ccris 3428, 2-4-pyridyl pyridine, 4-pyridyl-2-pyridine
PubChem CID 68488
IUPAC Name 2-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=NC=C2

Specifications

Melting Point 59°C to 63°C
Boiling Point 281°C
Quantity 1 g
UN Number UN2811
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 2,4'-Bipyridine
compliance-icons
Product Identifier
  • 2,4'-Bipyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Bipyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.