Learn More
2,4,6-Tris(difluoromethyl)-1,3,5-triazine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Tris(difluoromethyl)-1,3,5-triazine |
| CAS | 369-22-2 |
| Molecular Formula | C6H3N3F6 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H3F6N3/c7-1(8)4-13-5(2(9)10)15-6(14-4)3(11)12/h1-3H |
| SMILES | FC(F)c1nc(nc(n1)C(F)F)C(F)F |
| IUPAC Name | 2,4,6-tris(difluoromethyl)-1,3,5-triazine |
| Molecular Weight (g/mol) | 231.09 |
| Formula Weight | 231.0231 |