missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylaniline, 98%

Product Code. 11446676
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11446676

Brand: Thermo Scientific Alfa Aesar B24871.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6864-20-6 | C24H19N | 321.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6864-20-6
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.423
MDL Number MFCD00012371
InChI Key AGWKGKUGJDMKMT-UHFFFAOYSA-N
Synonym 2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
PubChem CID 629425
IUPAC Name 2,4,6-triphenylaniline
SMILES C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Formula Weight 321.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Triphenylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.