Learn More
2,4,6-Triphenylaniline, 98%
Product Code. 11466676
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11466676
Brand: Thermo Scientific Alfa Aesar B24871.14
Request Bulk or Custom Format
CAS: 6864-20-6 | C24H19N | 321.423 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6864-20-6
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.423
MDL Number MFCD00012371
InChI Key AGWKGKUGJDMKMT-UHFFFAOYSA-N
Synonym 2,4,6-triphenyl-anilin, 2,4,6-triphenylphenylamine, bidd:gt0713, 2,4,6-triphenylaniline, 3,5-diphenyl-1,1'-biphenyl-2-amine, 1,1':3',1-terphenyl-2'-amine,5'-phenyl, 5'-phenyl-1,1':3',1-terphenyl-2'-amine, 1,1':3',1-terphenyl-2'-amine, 5'-phenyl
PubChem CID 629425
IUPAC Name 2,4,6-triphenylaniline
SMILES C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
Specifications
Melting Point 122°C to 125°C
Quantity 25 g
Formula Weight 321.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Triphenylaniline