missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triiodoresorcinol, TRC

Product Code. 30451625
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451625 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451625 Supplier Toronto Research Chemicals Supplier No. TRCT79531525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Triiodoresorcinol
CAS 19403-92-0
Molecular Formula C6H3I3O2
Quantity 25 mg
Synonym 2,4,6-Triiodo-1,3-benzenediol
InChI Formula InChI=1S/C6H3I3O2/c7-2-1-3(8)6(11)4(9)5(2)10/h1,10-11H
SMILES Oc1c(I)cc(I)c(O)c1I
IUPAC Name 2,4,6-triiodobenzene-1,3-diol
Molecular Weight (g/mol) 487.8
Formula Weight 487.7267
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.