missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloroanisole D5 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317225
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317225 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317225 Supplier Dr. Ehrenstorfer Supplier No. DREA17714601ME100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Trichloroanisole D5
CAS 352439-08-8
Solvent or Matrix Methanol
Molecular Formula C7 2H5 Cl3 O
Quantity 1 mL
InChI Formula InChI=1S/C7H5Cl3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3/i1D3,2D,3D
SMILES [2H]c1c(Cl)c([2H])c(Cl)c(OC([2H])([2H])[2H])c1Cl
IUPAC Name 1,3,5-trichloro-2,4-dideuterio-6-(trideuteriomethoxy)benzene
Molecular Weight (g/mol) 216.5
Formula Weight 214.972 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.