Learn More
2,4,6-Tribromophenyl Acetate, TRC
Product Code. 30456921
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456921 1 g 1g
30451097 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30456921 Supplier Toronto Research Chemicals Supplier No. TRCT7726251G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 607-95-4
Molecular Formula C8 H5 Br3 O2
Molecular Weight (g/mol) 372.84
Synonym 2,4,6-Tribromo-phenol 1-Acetate, NSC 404079
IUPAC Name (2,4,6-tribromophenyl) acetate
SMILES CC(=O)Oc1c(Br)cc(Br)cc1Br
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H5Br3O2/c1-4(12)13-8-6(10)2-5(9)3-7(8)11/h2-3H,1H3
Formula Weight 369.784
Chemical Name or Material 2,4,6-Tribromophenyl Acetate