Learn More
2,4,5-Trichloronitrobenzene- 13C6, TRC
Product Code. 30457100
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.50mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457100 2.5 mg 2.5mg
30456635 0.5 mg 0.50mg
30452230 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30457100 Supplier Toronto Research Chemicals Supplier No. TRCT7743162.5MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4,5-Trichloronitrobenzene- 13C6
Molecular Formula 13C6 H2 Cl3 N O2
Quantity 2.5 mg
Synonym 1,2,4-Trichloro-5-nitrobenzene-13C6, 1,4,5-Trichloro-2-nitrobenzene-13 C6, 2,4,5-Trichloro-1-nitrobenzene- 13C6, 3,4,6-Trichloronitrobenzene- 13 C6
InChI Formula InChI=1S/C6H2Cl3NO2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H/i1+1,2+1,3+1,4+1,5+1,6+1
SMILES [O-][N+](=O)[13c]1[13cH][13c](Cl)[13c](Cl)[13cH][13c]1Cl
IUPAC Name 1,2,4-trichloro-5-nitro(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene
Molecular Weight (g/mol) 232.4
Formula Weight 230.935