missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%

Product Code. p-7026153
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499457 25 g 25g
11409467 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499457 Supplier Thermo Scientific Alfa Aesar Supplier No. B21989.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35011-47-3 | C4H9N5O5S | 239.206 g/mol

2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4H9N5O5S
Molecular Weight (g/mol) 239.206
MDL Number MFCD00012785
InChI Key RSKNEEODWFLVFF-UHFFFAOYSA-N
Synonym 2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
PubChem CID 296856
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 4621048
Solubility Information Soluble in DMSO.
Formula Weight 239.21
Percent Purity 97%
Chemical Name or Material 2,4,5-Triamino-6-hydroxypyrimidine sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.