missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%

Product Code. 11409467
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11409467

Brand: Thermo Scientific Alfa Aesar B21989.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35011-47-3 | C4H9N5O5S | 239.206 g/mol

2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4H9N5O5S
Molecular Weight (g/mol) 239.206
MDL Number MFCD00012785
InChI Key RSKNEEODWFLVFF-UHFFFAOYSA-N
Synonym 2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
PubChem CID 296856
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 4621048
Solubility Information Soluble in DMSO.
Formula Weight 239.21
Percent Purity 97%
Chemical Name or Material 2,4,5-Triamino-6-hydroxypyrimidine sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.