Learn More
2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol, 97%, Thermo Scientific™
Product Code. 15440739
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440739 1 g 1g
15428546 250 mg 250mg
15450739 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15440739 Supplier Thermo Scientific Alfa Aesar Supplier No. H63830.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5521-38-0
Molecular Formula C12H17N3O3
Molecular Weight (g/mol) 251.286
MDL Number MFCD00511837
InChI Key WVNPWSRYQDRSIS-UHFFFAOYSA-N
Synonym 2-4-4-nitrophenyl piperazin-1-yl ethanol, 2-4-4-nitrophenyl piperazino-1-ethanol, 2-4-4-nitrophenyl piperazin-1-yl ethan-1-ol, 2-4-4-nitro-phenyl-piperazin-1-yl-ethanol, 2-4-4-nitrophenyl piperazinyl ethan-1-ol, nitrophenylpiperazinoethanol, cambridge id 5139370, 4-4-nitrophenyl-1-piperazine-ethanol, 2-4-4-nitrophenyl-1-piperazinyl ethanol
PubChem CID 2763936
IUPAC Name 2-[4-(4-nitrophenyl)piperazin-1-yl]ethanol
SMILES C1CN(CCN1CCO)C2=CC=C(C=C2)[N+](=O)[O-]
Specifications
Quantity 1 g
Formula Weight 251.28
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-[4-(4-Nitrophenyl)-1-piperazinyl]ethanol