Learn More
2-(3-Fluorophenyl)pyrrolidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(3-Fluorophenyl)pyrrolidine |
| CAS | 298690-72-9 |
| Molecular Formula | C10H12FN |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C10H12FN/c11-9-4-1-3-8(7-9)10-5-2-6-12-10/h1,3-4,7,10,12H,2,5-6H2 |
| SMILES | Fc1cccc(c1)C2CCCN2 |
| Molecular Weight (g/mol) | 165.21 |
| Formula Weight | 165.0954 |