Learn More
2-(3-Fluorophenoxy)ethylamine hydrochloride, 98%, Thermo Scientific™
Product Code. 15449255
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15449255
Brand: Thermo Scientific Alfa Aesar H51681.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 372183-68-1
Molecular Formula C8H10FNO
Molecular Weight (g/mol) 155.172
MDL Number MFCD11099553
InChI Key UTSSEDORJLCBMS-UHFFFAOYSA-N
Synonym 2-3-fluorophenoxy ethylamine, 2-3-fluorophenoxy ethanamine, 2-3-fluoro-phenoxy-ethylamine, 1-2-aminoethoxy-3-fluorobenzene, ethanamine,2-3-fluorophenoxy, 2-3-fluorophenoxy ethan-1-amine, ethanamine, 2-3-fluorophenoxy, pubchem19158, acmc-1cfl9
PubChem CID 14245330
IUPAC Name 2-(3-fluorophenoxy)ethanamine
SMILES C1=CC(=CC(=C1)F)OCCN
Specifications
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 191.63
Percent Purity 98%
Chemical Name or Material 2-(3-Fluorophenoxy)ethylamine hydrochloride