Learn More
2,3-Dimethylfuran, 99%
Product Code. 11342747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342747 1 g 1g
11352747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342747 Supplier Thermo Scientific Alfa Aesar Supplier No. L15705.03
Request Bulk or Custom Format
CAS: 14920-89-9 | C6H8O | 96.13 g/mol
2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethylfuran may be used in the synthesis of 2,9-dioxabicyclo[3.3.1]nonane. Ireland Alcohol was synthesized in from 2,3-dimethylfuran.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14920-89-9
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00153893
InChI Key FJSKXQVRKZTKSI-UHFFFAOYSA-N
Synonym furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a
PubChem CID 34337
IUPAC Name 2,3-dimethylfuran
SMILES CC1=C(C)C=CO1
Specifications
Boiling Point 42°C (15 mmHg)
Flash Point −1°C (30°F)
Quantity 1 g
UN Number UN1992
Beilstein 104572
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylfuran