Learn More
2,3-Dideoxy-2-methylene-D-glycero-D-galacto-nononic Acid Gamma-Lactone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Dideoxy-2-methylene-D-glycero-D-galacto-nononic Acid gamma-L |
| CAS | 289697-66-1 |
| Molecular Formula | C10 H16 O7 |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C10H16O7/c1-4-2-6(17-10(4)16)8(14)9(15)7(13)5(12)3-11/h5-9,11-15H,1-3H2/t5-,6+,7-,8-,9+/m1/s1 |
| SMILES | OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)[C@@H]1CC(=C)C(=O)O1 |
| IUPAC Name | (5S)-3-methylidene-5-[(1S,2S,3R,4R)-1,2,3,4,5-pentahydroxypentyl]oxolan-2-one |
| Molecular Weight (g/mol) | 248.23 |
| Formula Weight | 248.0896 |