missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenoxyacetic Acid, TRC

Product Code. 30373592
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30373592

Brand: Toronto Research Chemicals TRCD4741535G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dichlorophenoxyacetic Acid
CAS 2976-74-1
Molecular Formula C8 H6 Cl2 O3
Quantity 5 g
Synonym Acetic acid, (2,3-dichlorophenoxy)- (8CI,9CI), 2-(2,3-Dichlorophenoxy)acetic acid, 2,3-D, 2,3-Dichlorophenoxyacetic acid, NSC 74462
InChI Formula InChI=1S/C8H6Cl2O3/c9-5-2-1-3-6(8(5)10)13-4-7(11)12/h1-3H,4H2,(H,11,12)
SMILES OC(=O)COc1cccc(Cl)c1Cl
IUPAC Name 2-(2,3-dichlorophenoxy)acetic acid
Molecular Weight (g/mol) 221.04
Formula Weight 219.9694
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.