missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorophenoxyacetic Acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 2,3-Dichlorophenoxyacetic Acid |
| CAS | 2976-74-1 |
| Molecular Formula | C8 H6 Cl2 O3 |
| Quantity | 25 g |
| Synonym | Acetic acid, (2,3-dichlorophenoxy)- (8CI,9CI), 2-(2,3-Dichlorophenoxy)acetic acid, 2,3-D, 2,3-Dichlorophenoxyacetic acid, NSC 74462 |
| InChI Formula | InChI=1S/C8H6Cl2O3/c9-5-2-1-3-6(8(5)10)13-4-7(11)12/h1-3H,4H2,(H,11,12) |
| SMILES | OC(=O)COc1cccc(Cl)c1Cl |
| IUPAC Name | 2-(2,3-dichlorophenoxy)acetic acid |
| Molecular Weight (g/mol) | 221.04 |
| Formula Weight | 219.9694 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction