missing translation for 'onlineSavingsMsg'
Learn More

2,3’-Anhydrothymidine, TRC

Product Code. 30336272
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336272 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336272 Supplier Toronto Research Chemicals Supplier No. TRCA656500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3'-Anhydrothymidine
CAS 15981-92-7
Molecular Formula C10H12N2O4
Quantity 250 mg
Synonym 2,5-Methano-5H,9H-pyrimido[2,1-b][1,5,3]dioxazepin-9-one, 2,3-dihydro-3-(hydroxymethyl)-8-methyl-, (2R,3R,5R)-, 2,5-Methano-5H,9H-pyrimido[2,1-b][1,5,3]dioxazepin-9-one, 2,3-dihydro-3-(hydroxymethyl)-8-methyl- (7CI), 2,5-Methano-5H,9H-pyrimido[2,1-b][1,5,3]dioxazepin-9-one, 2,3-dihydro-3-(hydroxymethyl)-8-methyl-, [2R-(2α,3β,5α)]-, 2,5-Methano-5H,9H-pyrimido[2,1-b][1,5,3]dioxazepin-9-one, 2,3-dihydro-3-(hydroxymethyl)-8-methyl-, stereoisomer (8CI), (2R,3R,5R)-2,3-Dihydro-3-(hydroxymethyl)-8-methyl-2,5-methano-5H,9H-pyrimido[2,1-b][1,5,3]dioxazepin-9-one, 2,3'-Anhydro-1-(2'-deoxy-β-D-threo-pentofuranosyl)thymine, 2,3'-O-Cyclothymidine, NSC 144601, O2,3'-Cyclothymidine
InChI Formula InChI=1S/C10H12N2O4/c1-5-3-12-8-2-6(7(4-13)15-8)16-10(12)11-9(5)14/h3,6-8,13H,2,4H2,1H3
SMILES CC1=CN2[C@H]3C[C@@H](OC2=NC1=O)[C@@H](CO)O3
IUPAC Name 10-(hydroxymethyl)-4-methyl-8,11-dioxa-2,6-diazatricyclo[7.2.1.02,7]dodeca-3,6-dien-5-one
Molecular Weight (g/mol) 224.21
Formula Weight 224.0797
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.