Learn More
2,3,5-Trichloropyridin-4-amine, TRC
Product Code. 30453861
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453861 50 mg 50mg
30450130 100 mg -
30452193 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30453861 Supplier Toronto Research Chemicals Supplier No. TRCT89959050MG
 Restricted Item
Estimated Shipment: 10-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28443-69-8
Molecular Formula C5 H3 Cl3 N2
Molecular Weight (g/mol) 197.45
Synonym Picloram-decarboxy
IUPAC Name 2,3,5-trichloropyridin-4-amine
SMILES Nc1c(Cl)cnc(Cl)c1Cl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C5H3Cl3N2/c6-2-1-10-5(8)3(7)4(2)9/h1H,(H2,9,10)
Formula Weight 195.9362
Chemical Name or Material Picloram-decarboxy