Learn More
2,3,5,6-Tetrachloropyridine, 98%
Product Code. 11358776
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11358776
Brand: Thermo Scientific Alfa Aesar L19458.14
Request Bulk or Custom Format
CAS: 2402-79-1 | C5HCl4N | 216.87 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2402-79-1
Molecular Formula C5HCl4N
Molecular Weight (g/mol) 216.87
MDL Number MFCD00092428
InChI Key FATBKZJZAHWCSL-UHFFFAOYSA-N
Synonym pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine
PubChem CID 16990
IUPAC Name 2,3,5,6-tetrachloropyridine
SMILES ClC1=CC(Cl)=C(Cl)N=C1Cl
Specifications
Melting Point 89°C to 93°C
Boiling Point 250°C to 252°C
Flash Point 188°C (370°F)
Quantity 25 g
UN Number UN3077
Beilstein 129639
Formula Weight 216.88
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrachloropyridine