missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic Acid, TRC

Product Code. 30456764
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456764 500 mg -
30454596 5 g 5g
30451160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456764 Supplier Toronto Research Chemicals Supplier No. TRCT888018500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-02-6
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.11
Synonym 4-Pyrogallolcarboxylic Acid, NSC 27436, 2,3,4-Trihydroxybenzenecarboxylic Acid
IUPAC Name 2,3,4-trihydroxybenzoic acid
SMILES OC(=O)c1ccc(O)c(O)c1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H6O5/c8-4-2-1-3(7(11)12)5(9)6(4)10/h1-2,8-10H,(H,11,12)
Formula Weight 170.0215
Chemical Name or Material 2,3,4-Trihydroxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.