Learn More
2,3,4-Trihydroxybenzaldehyde, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,3,4-Trihydroxybenzaldehyde |
| CAS | 2144-08-3 |
| Molecular Formula | C7H6O4 |
| Quantity | 5 g |
| Synonym | 3,4-Dihydroxysalicylaldehyde, NSC 22595 |
| InChI Formula | InChI=1S/C7H6O4/c8-3-4-1-2-5(9)7(11)6(4)10/h1-3,9-11H |
| SMILES | Oc1ccc(C=O)c(O)c1O |
| IUPAC Name | 2,3,4-trihydroxybenzaldehyde |
| Molecular Weight (g/mol) | 154.12 |
| Formula Weight | 154.0266 |