Learn More
2-(3,4-Dihydroxyphenyl)ethyl-2,2-d2-amine HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Dopamine-D2 Hydrochloride |
| CAS | 27160-01-6 |
| Molecular Formula | C8 D2 H9 N O2 . Cl H |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzenediol, 4-(2-aminoethyl-1,1-d2)-, hydrochloride (9 CI), Pyrocatechol, 4-(2-aminoethyl-1,1-d2)-, hydrochloride (8 CI), Dopamine-d2 hydrochloride, Dopamine-D2 (2-aminoethyl-1,1-D2) hydrochloride, 4-(2-Aminoethyl-1,1-d2)-1,2-benzenediol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethyl-2,2-d2-amine HCl |
| InChI Formula | InChI=1 S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11 H,3-4,9H2;1 H/i3D2; |
| SMILES | Cl.[2 H]C([2 H])(CN)c1ccc(O)c(O)c1 |
| IUPAC Name | 4-(2-amino-1,1-dideuterioethyl)benzene-1,2-diol;hydrochloride |
| Molecular Weight (g/mol) | 191.652 |
| Formula Weight | 191.068 g/mol |
| Show More |