Learn More
2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d4-amine HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Dopamine-D4 Hydrochloride |
| CAS | 203633-19-6 |
| Molecular Formula | C8 2H4 H7 N O2 . Cl H |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11 H,3-4,9H2;1 H/i3D2,4D2; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])c1ccc(O)c(O)c1 |
| IUPAC Name | 4-(2-amino-1,1,2,2-tetradeuterioethyl)benzene-1,2-diol;hydrochloride |
| Molecular Weight (g/mol) | 193.66 |
| Formula Weight | 193.0808 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |