Learn More
2,3,4,6-Tetra-O-benzyl-1-deoxynojirimycin Hydrochloric Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4,6-Tetra-O-benzyl-1-deoxynojirimycin Hydrochloric Acid Salt |
| CAS | 72983-76-7 |
| Molecular Formula | C34H37NO4 . HCl |
| Quantity | 25 mg |
| Synonym | (2R,3R,4R,5S)-3,4,5-Tris(phenylmethoxy)-2-[(phenylmethoxy)methyl]piperidine Hydrochloride |
| InChI Formula | InChI=1S/C34H37NO4.ClH/c1-5-13-27(14-6-1)22-36-26-31-33(38-24-29-17-9-3-10-18-29)34(39-25-30-19-11-4-12-20-30)32(21-35-31)37-23-28-15-7-2-8-16-28;/h1-20,31-35H,21-26H2;1H/t31-,32+,33-,34-;/m1./s1 |
| SMILES | Cl.C(OCc1ccccc1)[C@H]2NC[C@H](OCc3ccccc3)[C@@H](OCc4ccccc4)[C@@H]2OCc5ccccc5 |
| IUPAC Name | (2R,3R,4R,5S)-3,4,5-tris(phenylmethoxy)-2-(phenylmethoxymethyl)piperidine;hydrochloride |
| Molecular Weight (g/mol) | 560.1 |
| Formula Weight | 559.2489 |