Learn More
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl bromide, 95%, stabilized
Product Code. 10088761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10088761 10 g 10g
10232612 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10088761 Supplier Thermo Scientific Acros Supplier No. 353690100
Request Bulk or Custom Format
CAS: 3068-32-4 | C14H19BrO9 | 411.201 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3068-32-4 , 471-34-1
Molecular Formula C14H19BrO9
MDL Number MFCD00063686
Synonym 5r,6r-3,4,5-triacetyloxy-6-bromanyl-oxan-2-yl methyl ethanoate, acetic acid 5r,6r-3,4,5-triacetyloxy-6-bromo-2-oxanyl methyl ester, d-alpha-tetraacetobromogalactose
PubChem CID 17750873
Specifications
CAS Min % 94.0
CAS Max % 100.0
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Specific Rotation Condition +210.00 (20.00°C=3,CHCl3)
Specific Rotation +200° to +220° (20°C, 589 nm) (c=3, CHCl3)
InChI Key CYAYKKUWALRRPA-UHFFFAOYNA-N
SMILES CC(=O)OCC1OC(Br)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
IUPAC Name [3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
Stabilizer 2% CaCO3
Molecular Weight (g/mol) 411.20
Formula Weight 411.21
Percent Purity ≥94%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4,6-Tetra-O-acetyl-a-D-galactopyranosyl bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.