Learn More
2,3,4,5,6-Pentafluorophenol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 771-61-9 |
| Molecular Formula | C6HF5O |
| MDL Number | MFCD00002156 |
| Quantity | 100 g |
| SMILES | OC1=C(F)C(F)=C(F)C(F)=C1F |
| Molecular Weight (g/mol) | 184.06 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,3,4,5,6-Pentafluorophenol |