missing translation for 'onlineSavingsMsg'
Learn More

2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol-d12 (Major), TRC

Product Code. 30336270
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336270 5 mg 5mg
30343406 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30336270 Supplier Toronto Research Chemicals Supplier No. TRCB2070475MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol-d12 (Major)
Purity Grade Notes HPLC
Molecular Formula C22 D12 H17 N3 O
Quantity 5 mg
Synonym 2-(2-Hydroxy-3,5-di-tert-amylphenyl)-2H-benzotriazole-d12, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)benzotriazole-d12, 2-(2-Hydroxy-3,5-di-tert-pentylphenyl)benzotriazole-d12, 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylpropyl)phenol-d12, 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol-d12, 2-(2'-Hydroxy-3',5'-di-tert-amylphenyl)benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)-2H-benzotriazole-d12, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole-d12, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)-2H-benzotriazole-d12, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)benzotriazole-d12, 2-(3',5'-Di-tert-amyl-2'-hydroxyphenyl)benzotriazole-d12, 2-[2-Hydroxy-3,5-bis(1,1-dimethylpropyl)phenyl]-2H-benzotriazole-d12, Chisorb 328-d12, Cyasorb UV 2337-d12, Eversorb 74-d12, Kemisorb 74-d12, Light Stabilizer 328-d12, Lowilite 28-d12, Seesorb 704-d12, Sumisorb 350-d12, THUV 328-d12, Tin 328-d12, Tinuvin 328-d12, Tinuvin PA 238-d12, UV 2337-d12, UV 328-d12, UV 350-d12, UV 74-d12, Viosorb 591-d12
InChI Formula InChI=1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3/i3D3,4D3,5D3,6D3
SMILES [2H]C([2H])([2H])C(CC)(c1cc(c(O)c(c1)C(CC)(C([2H])([2H])[2H])C([2H])([2H])[2H])n2nc3ccccc3n2)C([2H])([2H])[2H]
IUPAC Name 2-(benzotriazol-2-yl)-4,6-bis[1,1,1-trideuterio-2-(trideuteriomethyl)butan-2-yl]phenol
Molecular Weight (g/mol) 363.559
Formula Weight 363.306
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.