Learn More
2-(2-Piperidinoethoxy)ethanol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Piperidinoethoxy)ethanol |
| CAS | 3603-43-8 |
| Molecular Formula | C9H19NO2 |
| Quantity | 1 g |
| Synonym | 2-[2-(1-Piperidinyl)ethoxy]-ethanol, 2-[2-(1-Piperidinyl)ethoxy]ethanol |
| InChI Formula | InChI=1S/C9H19NO2/c11-7-9-12-8-6-10-4-2-1-3-5-10/h11H,1-9H2 |
| SMILES | CC1=NC2=CC([N+]([O-])=O)=C([N+]([O-])=O)C=C2N1 |
| IUPAC Name | 2-(2-(piperidin-1-yl)ethoxy)ethan-1-ol |
| Molecular Weight (g/mol) | 173.25 |
| Formula Weight | 173.14 |