missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Oxopiperazin-1-yl)acetic Acid Hydrochloride, TRC

Product Code. 30427019
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427019 50 mg -
30410185 100 mg -
30413935 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427019 Supplier Toronto Research Chemicals Supplier No. TRCO85883050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32705-76-3
Molecular Formula C6H10N2O3 . HCl
Molecular Weight (g/mol) 194.61
Synonym 1-Piperazineacetic acid, 2-oxo-, hydrochloride (1:1) (ACI), 1-Piperazineacetic acid, 2-oxo-, monohydrochloride (8CI), 2-Oxo-1-piperazineacetic acid hydrochloride, 2-(2-Oxopiperazin-1-yl)acetic acid hydrochloride, 2-(2-Oxopiperazino)acetic acid hydrochloride, Ethylenediamine-N,N'-diacetic acid lactam hydrochloride
IUPAC Name 2-(2-oxopiperazin-1-yl)acetic acid;hydrochloride
SMILES Cl.OC(=O)CN1CCNCC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H10N2O3.ClH/c9-5-3-7-1-2-8(5)4-6(10)11;/h7H,1-4H2,(H,10,11);1H
Formula Weight 194.0458
Chemical Name or Material 2-(2-Oxopiperazin-1-yl)acetic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.