Learn More
2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 95%
Product Code. 15440107
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440107 250 mg 250mg
15430107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15440107 Supplier Thermo Scientific Alfa Aesar Supplier No. H66633.MD
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 104206-65-7 | C14H10F3NO5 | 329.231 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104206-65-7
Molecular Formula C14H10F3NO5
Molecular Weight (g/mol) 329.231
MDL Number MFCD01752192
InChI Key OUBCNLGXQFSTLU-UHFFFAOYSA-N
Synonym nitisinone, orfadin, nitisone, ntbc, 2-2-nitro-4-trifluoromethyl benzoyl cyclohexane-1,3-dione, 2-2-nitro-4-trifluoromethylbenzoyl-1,3-cyclohexanedione, nitisinone usan:inn, nitisinone inn, nitisinona
PubChem CID 115355
ChEBI CHEBI:50378
IUPAC Name 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione
SMILES C1CC(=O)C(C(=O)C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)[N+](=O)[O-]
Specifications
Quantity 250 mg
Formula Weight 329.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione