missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Naphthoxy)ethanol, 98+%

Product Code. 11404743
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11404743

Brand: Thermo Scientific Alfa Aesar A11636.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-20-9 | C12H12O2 | 188.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-20-9
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00016809
InChI Key BQPBZDSDFCDSAO-UHFFFAOYSA-N
Synonym 2-2-naphthyloxy ethanol, 2-2-naphthoxy ethanol, 2-naphthalen-2-yloxy ethanol, anavenol, 2-2-hydroxyethoxy naphthalene, ethanol, 2-2-naphthalenyloxy, 2-2-naphthalenyloxy ethanol, unii-7m7cut7ccu, ethylene glycol mono-2-naphthyl ether, .beta.-naphthoxyethanol
PubChem CID 7131
IUPAC Name 2-naphthalen-2-yloxyethanol
SMILES C1=CC=C2C=C(C=CC2=C1)OCCO

Specifications

Melting Point 72°C to 76°C
Quantity 25 g
Beilstein 2086973
Merck Index 14,6413
Formula Weight 188.23
Percent Purity ≥98%
Chemical Name or Material 2- (2-Naphthoxy)ethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.