missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile, 98%, Thermo Scientific™

Product Code. 15409125
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409125 1 g 1g
15419125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409125 Supplier Thermo Scientific Alfa Aesar Supplier No. H51040.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213749-91-8
Molecular Formula C19H26N2O2
Molecular Weight (g/mol) 314.429
MDL Number MFCD03701544
InChI Key YGAQISIJKKIHRN-UHFFFAOYSA-N
Synonym 2-2-ethylhexyloxy-5-methoxy-1,4-benzenediacetonitrile, 2,2'-2-2-ethylhexyloxy-5-methoxy-1,4-phenylene diacetonitrile, 2,2'-2-2-ethylhexyl oxy-5-methoxy-1,4-phenylene diacetonitrile, 2-4-cyanomethyl-5-2-ethylhexyl oxy-2-methoxyphenyl acetonitrile, 2-methoxy-5-2-ethylhexyloxy benzene-1, 2-methoxy-5-2'-ethylhexyloxy benzene-1, 1,4-bis cyanomethyl-2 2'-ethylhexyloxy-5-methoxybenzene, 2-4-cyanomethyl-2-2-ethylhexoxy-5-methoxyphenyl acetonitrile, 2-methoxy-5-2'-ethylhexyloxy benzene-1,4-diacetonitrile
PubChem CID 4115353
IUPAC Name 2-[4-(cyanomethyl)-2-(2-ethylhexoxy)-5-methoxyphenyl]acetonitrile
SMILES CCCCC(CC)COC1=CC(=C(C=C1CC#N)OC)CC#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 314.42
Percent Purity 98%
Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.