missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylcyclopropanecarboxylic Acid Ethyl Ester, TRC

Product Code. 30382497
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382497 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382497 Supplier Toronto Research Chemicals Supplier No. TRCD487520250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2-Diphenylcyclopropanecarboxylic Acid Ethyl Ester
CAS 37555-46-7
Molecular Formula C18 H18 O2
Quantity 250 mg
Synonym Ethyl 2,2-Diphenylcyclopropanecarboxylate, Ethyl 2,2-Diphenylcyclopropylcarboxylate
InChI Formula InChI=1S/C18H18O2/c1-2-20-17(19)16-13-18(16,14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12,16H,2,13H2,1H3
SMILES CCOC(=O)C1CC1(c2ccccc2)c3ccccc3
IUPAC Name ethyl 2,2-diphenylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 266.33
Formula Weight 266.1307
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.