Learn More
2,2-Dimethyloctanoic Acid, TRC
Product Code. 30377948
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377948 500 mg
30371147 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30377948 Supplier Toronto Research Chemicals Supplier No. TRCD476185500MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29662-90-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.26
Synonym 2,2-Dimethyloctanoic Acid, α,α-Dimethyloctanoic Acid, 2,2-Dimethyl-Octanoic Acid
IUPAC Name 2,2-dimethyloctanoic acid
SMILES O=C(O)C(C)(C)CCCCCC
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H20O2/c1-4-5-6-7-8-10(2,3)9(11)12/h4-8H2,1-3H3,(H,11,12)
Formula Weight 172.1463
Chemical Name or Material 2,2-Dimethyloctanoic Acid