Learn More
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 97%, Thermo Scientific™
Product Code. p-2696630
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 10783043
Brand: Thermo Scientific Maybridge CC27201DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 42327-95-7
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD03426826
InChI Key AVVACPBRCFXZMR-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carboxylic acid, 2, 3-dihydro-2, 2-dimethylbenzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carboxylic acid
PubChem CID 2795469
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-carboxylic acid
SMILES CC1(CC2=CC=CC(=C2O1)C(=O)O)C
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carboxylic acid