Learn More
2,2-Difluoropropionic acid, 97%, Thermo Scientific Chemicals
Product Code. p-7028636
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15433347
Brand: Thermo Scientific Alfa Aesar H26030.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 373-96-6
Molecular Formula C3H4F2O2
Molecular Weight (g/mol) 110.06
MDL Number MFCD03093764
InChI Key PMWGIVRHUIAIII-UHFFFAOYSA-N
Synonym 2,2-difluoropropionic acid, 2,2-difluoropropionicacid, 2,2-difluoro-propionic acid, propanoic acid, 2,2-difluoro, acmc-1ctsu, 2,2-difluorpropionic acid
PubChem CID 2782825
IUPAC Name 2,2-difluoropropanoic acid
SMILES CC(F)(F)C(O)=O
Specifications
Melting Point 36°C to 44°C
Boiling Point 142°C
Flash Point 66°C (150°F)
Linear Formula CH3CF2COOH
Quantity 250 mg
UN Number UN3261
Formula Weight 110.06
Percent Purity 97%
Chemical Name or Material 2,2-Difluoropropionic acid