missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dichloro-4,4'-bipyridine, 95%

Product Code. 11322799
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322799 500 mg 500mg
11312799 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322799 Supplier Thermo Scientific Alfa Aesar Supplier No. H27695.ME
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 53344-74-4 | C10H6Cl2N2 | 225.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53344-74-4
Molecular Formula C10H6Cl2N2
Molecular Weight (g/mol) 225.072
MDL Number MFCD01106345
InChI Key KSELKNWXBOXHEV-UHFFFAOYSA-N
Synonym 2,2'-dichloro-4,4'-bipyridine, 2-chloro-4-2-chloropyridin-4-yl pyridine, 2,2'-dichloro-4,4'bipyridine, 2,2-dichloro-4,4-bipyridine, pubchem19451, bpcl37, acmc-1ap45, 4,4'-bi 2-chloropyridine, 2,2'-dichloro-4,4' bipyridine
PubChem CID 5172097
IUPAC Name 2-chloro-4-(2-chloropyridin-4-yl)pyridine
SMILES C1=CN=C(C=C1C2=CC(=NC=C2)Cl)Cl

Specifications

Melting Point 240°C to 243°C
Quantity 500 mg
Formula Weight 225.08
Percent Purity 95%
Chemical Name or Material 2,2'-Dichloro-4,4'-bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.