Learn More
2-(2-Chloro-4-iodophenylamino)-3,4-difluorobenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Chloro-4-iodophenylamino)-3,4-difluorobenzoic Acid |
| CAS | 303175-44-2 |
| Molecular Formula | C13 H7 Cl F2 I N O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C13H7ClF2INO2/c14-8-5-6(17)1-4-10(8)18-12-7(13(19)20)2-3-9(15)11(12)16/h1-5,18H,(H,19,20) |
| SMILES | OC(=O)c1ccc(F)c(F)c1Nc2ccc(I)cc2Cl |
| IUPAC Name | 2-(2-chloro-4-iodoanilino)-3,4-difluorobenzoic acid |
| Molecular Weight (g/mol) | 409.55 |
| Formula Weight | 408.9178 |