Learn More
2-(2-Bromoacetamide)-3-benzoyl-4,5,6,7-tetrahydrobenzo[b]thiophene, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Bromoacetamide)-3-benzoyl-4,5,6,7-tetrahydrobenzo[b]thiophene |
| CAS | 29462-27-9 |
| Molecular Formula | C17H16BrNO2S |
| Quantity | 50 mg |
| Synonym | N-(3-benzoyl-4,5,6,7-tetrahydrobenzo[b]thien-2-yl)-2-bromoacetamide |
| InChI Formula | InChI=1S/C17H16BrNO2S/c18-10-14(20)19-17-15(12-8-4-5-9-13(12)22-17)16(21)11-6-2-1-3-7-11/h1-3,6-7H,4-5,8-10H2,(H,19,20) |
| SMILES | BrCC(=O)Nc1sc2CCCCc2c1C(=O)c3ccccc3 |
| IUPAC Name | N-(3-benzoyl-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)-2-bromoacetamide |
| Molecular Weight (g/mol) | 378.28 |
| Formula Weight | 377.0085 |