Learn More
2-(2-Bromo-6-chlorophenyl)acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Bromo-6-chlorophenyl)acetic Acid |
| CAS | 37777-77-8 |
| Molecular Formula | C8H6O2ClBr |
| Quantity | 10 mg |
| Synonym | 2-Chloro-6-bromophenylacetic Acid, 2-Bromo-6-chlorobenzeneacetic Acid |
| InChI Formula | InChI=1S/C8H6BrClO2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12) |
| SMILES | OC(=O)Cc1c(Cl)cccc1Br |
| IUPAC Name | 2-(2-bromo-6-chlorophenyl)acetic acid |
| Molecular Weight (g/mol) | 249.49 |
| Formula Weight | 247.924 |