Learn More
2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane, TRC
Product Code. 30138024
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138024 10 g 10g
30011329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138024 Supplier Toronto Research Chemicals Supplier No. TRCB44740010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2971-36-0
Molecular Formula C14 H11 Cl3 O2
Molecular Weight (g/mol) 317.59
Synonym 4,4'-(2,2,2-Trichloroethylidene)diphenol, 4,4'-Dihydroxydiphenyltrichloroethane, 2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane, Bisdemethylmethoxychlor, HPTE, 1,1,1-Trichloro-2,2-bis(4-hydroxyphenyl)ethane, Hydroxychlor, NSC 7045, p,p'-Hydroxy-DDT, 1,1-Bis(4-hydroxyphenyl)-2,2,2-trichloroethane, 1,1-Bis(p-hydroxyphenyl)-2,2,2-trichloroethane, 2,2,2-Trichloro-1,1-bis(4-hydroxyphenyl)ethane, α,α-Bis(4-hydroxyphenyl)-β,β,β-trichloroethane;
IUPAC Name 4-[2,2,2-trichloro-1-(4-hydroxyphenyl)ethyl]phenol
SMILES Oc1ccc(cc1)C(c2ccc(O)cc2)C(Cl)(Cl)Cl
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H11Cl3O2/c15-14(16,17)13(9-1-5-11(18)6-2-9)10-3-7-12(19)8-4-10/h1-8,13,18-19H
Formula Weight 315.9825
Percent Purity >95%
Chemical Name or Material 2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane
Grade HPLC