Learn More
2,2,7-Trimethyloctane-3,5-dione, 97%
Product Code. 11353626
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11353626
Brand: Thermo Scientific Alfa Aesar L14639.06
Request Bulk or Custom Format
CAS: 69725-37-7 | C11H20O2 | 184.279 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69725-37-7
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00511279
InChI Key KEBPOWLGOOTMPK-UHFFFAOYSA-N
PubChem CID 112295
IUPAC Name 2,2,7-trimethyloctane-3,5-dione
SMILES CC(C)CC(=O)CC(=O)C(C)(C)C
Specifications
Boiling Point 44°C (0.2 mmHg)
Refractive Index 1.458
Quantity 5 g
Synonym 2,2,7-trimethyl-3,5-octanedione, 3,5-octanedione, 2,2,7-trimethyl, 2,2 7-trimethyloctane-3,5-dione, 3,5-octanedione,2,2,7-trimethyl, 2,2,7-trimethyl-3,5-octanedione #, 2,2,7-trimethyl-3,5-octadione
Formula Weight 184.28
Percent Purity ≥98%
Chemical Name or Material 2,2,7-Trimethyloctane-3,5-dione