Learn More
[2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]carbamic Acid-d6 1,1-Dimethylethyl Ester, TRC
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | [2-[(2,6-Dimethylphenyl)amino]-2-oxoethyl]carbamic Acid-d6 1,1-Dimethylethyl Ester |
| Molecular Formula | C20 H16 O |
| Quantity | 2.5 mg |
| InChI Formula | InChI=1S/C20H16O/c21-19-13-11-18(12-14-19)20(17-9-5-2-6-10-17)15-16-7-3-1-4-8-16/h1-15,21H/b20-15- |
| SMILES | Oc1ccc(cc1)\C(=C/c2ccccc2)\c3ccccc3 |
| IUPAC Name | 4-[(Z)-1,2-diphenylethenyl]phenol |
| Molecular Weight (g/mol) | 272.34 |
| Formula Weight | 272.12 |