missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,6,6-Tetramethylpiperidine-N-oxyl-4-amino-4-carboxylic Acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2,2,6,6-Tetramethylpiperidine-N-oxyl-4-amino-4-carboxylic Ac |
| CAS | 15871-57-5 |
| Molecular Formula | C10 H19 N2 O3 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H19N2O3/c1-8(2)5-10(11,7(13)14)6-9(3,4)12(8)15/h5-6,11H2,1-4H3,(H,13,14) |
| SMILES | CC1(C)CC(N)(CC(C)(C)N1[O])C(=O)O |
| IUPAC Name | 4-amino-1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine-4-carboxylic acid |
| Molecular Weight (g/mol) | 215.27 |
| Formula Weight | 215.1396 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction