Learn More
2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%
Product Code. 10713531 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
Product Code. 10713531
Brand: Thermo Scientific Acros 301452500
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 15871-57-5 | C10H19N2O3 | 215.27 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15871-57-5
Molecular Formula C10H19N2O3
Molecular Weight (g/mol) 215.27
MDL Number MFCD00800575
Synonym toac, unii-xdn8h1xm17, 4-amino-4-carboxy-2,2,6,6-tetramethylpiperidin-1-yl oxidanyl, 2,2,6,6-tetramethylpiperidine-n-oxide-4-amino-4-carboxylic acid, xdn8h1xm17, 4-amino-4-carboxy-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-amino-1, tempo-4-amino-4-carboxylic acid, 1-piperidinyloxy,4-amino-4-carboxy-2,2,6,6-tetramethyl, 4-amino-4-carboxy-2,2,6,6-tetramethylpiperidine 1-oxyl
Specifications
CAS Min % 95.0
CAS Max % 100.0
Melting Point 230°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15,9645
Formula Weight 215.27
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine-1-oxyl-4-amino-4-carboxylic acid, 96%