missing translation for 'onlineSavingsMsg'
Learn More

2,2',4,5,5'-Pentachlorobiphenyl, TRC

Product Code. 30428102
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428102 250 mg 250mg
30423857 10 mg 10mg
30416970 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428102 Supplier Toronto Research Chemicals Supplier No. TRCP220400250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37680-73-2
Molecular Formula C12 H5 Cl5
Molecular Weight (g/mol) 326.43
Synonym 1,1'-Biphenyl, 2,2',4,5,5'-pentachloro-, 2,2',4,5,5'-Pentachloro-1,1'-biphenyl, 2,2',4,5,5'-PCB, 2,2',4,5,5'-Pentachlorobiphenyl, 2,2',4',5,5'-Pentachlorobiphenyl, 2,4,5,2',5'-Pentachlorobiphenyl, 2,5,2',4',5'-Pentachlorobiphenyl, CB 101, K 101, PCB 101, 2,2',4,5,5'-PentaCB
IUPAC Name 1,2,4-trichloro-5-(2,5-dichlorophenyl)benzene
SMILES Clc1ccc(Cl)c(c1)c2cc(Cl)c(Cl)cc2Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H5Cl5/c13-6-1-2-9(14)7(3-6)8-4-11(16)12(17)5-10(8)15/h1-5H
Formula Weight 323.8834
Chemical Name or Material PCB No. 101
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.