Learn More
2,2',4,4',6,6'-Hexabromo-bibenzyl, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2',4,4',6,6'-Hexabromo-bibenzyl |
| CAS | 97759-27-8 |
| Molecular Formula | C14H8Br6 |
| Quantity | 50 mg |
| Synonym | 1,2-Bis(2,4,6-tribromophenyl)ethane, 1,1'-(1,2-Ethanediyl)bis[2,4,6-tribromobenzene] |
| InChI Formula | InChI=1S/C14H8Br6/c15-7-3-11(17)9(12(18)4-7)1-2-10-13(19)5-8(16)6-14(10)20/h3-6H,1-2H2 |
| SMILES | Brc1cc(Br)c(CCc2c(Br)cc(Br)cc2Br)c(Br)c1 |
| IUPAC Name | 1,3,5-tribromo-2-[2-(2,4,6-tribromophenyl)ethyl]benzene |
| Molecular Weight (g/mol) | 655.64 |
| Formula Weight | 649.5726 |