Learn More
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Henicosafluoroundecanoic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2058-94-8 |
| Molecular Formula | C11HF21O2 |
| MDL Number | MFCD00153268 |
| Quantity | 5 g |
| SMILES | O=C(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| Molecular Weight (g/mol) | 564.09 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Henicosafluoroundecanoic acid |